Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Pierce™ Polyacrylamide Spin Desalting Columns, 7K MWCO, 0.7 mL

Catalog No. PI89849
Change view
Click to view available options
Quantity:
25 Columns
50 Columns
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI89849 25 Columns
PI89862 50 Columns
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PI89849 Supplier Thermo Scientific™ Supplier No. 89849
Add to Cart
Add to Cart

Separate proteins and other macromolecules from low-molecular weight buffer salts and reagents with Thermo Scientific Pierce 7K MWCO (molecular-weight cutoff) Polyacrylamide Spin Desalting Columns, which are porous polyacrylamide-based gel-filtration columns for biological sample preparation.

Desalting and buffer exchange are two of the most widely used applications in resin filtration chromatography. Thermo Scientific Pierce Polyacrylamide Spin Desalting Columns, 7K MWCO (molecular-weight cutoff), are ready-to-use, disposable, gel-filtration columns for the separation of proteins and other macromolecules from low-molecular-weight buffer salts and reagents.

Pierce Polyacrylamide Spin Desalting Columns, 7K MWCO, are pre-packed with porous polyacrylamide beads and enable buffer-exchange and desalting of proteins and other macromolecular samples based on a molecular-weight cutoff (MWCO) of 7000 Da. Unlike other size-exclusion media, polyacrylamide resin is not subject to enzymatic degradation and will not serve as a nutrient for microbial growth. The resin is very hydrophilic, thereby minimizing undesirable binding interactions between the resin and sample molecules. Contamination with low molecular weight sugars (which may occur with crosslinked dextran resin) is not a concern when using polyacrylamide resin. Oxidizing agents can be removed without destroying the support. This resin is susceptible to hydrolysis of amide groups under extreme pH conditions, so an operating pH of 2–10 is recommended at room temperature. The polyacrylamide resin can also be autoclaved at pH 5.5–6.5 for 30 minutes at 120°C.

The columns use a resin with beads that have a wet diameter of 45 to 90 μM and are ideal for separating peptides and small macromolecules (>7 kDa) from buffer salts and other compounds (less than 1K MWCO).

Features of polyacrylamide resin include:
• Stable in water, salt solutions, organic solvents, and alkaline or acidic conditions
• Excellent flow properties
• Heat stable

Applications
• Removing salts from protein solutions
• Removing phenol from nucleic acid preparations
• Separating excess crosslinker from conjugate preparations
• Removing excess derivatizing agents from modified proteins
• Removing unreacted dye from fluorescent antibodies
• Removing free radiolabel from labeled proteins
• Buffer exchange

TRUSTED_SUSTAINABILITY

Specifications

Description Pierce™ Polyacrylamide Spin Desalting Columns, 7K MWCO, 0.7 mL
Product Type Spin Desalting Column
Content And Storage Upon receipt store at 4°C.
Format Spin Column
Purification Target Buffer Exchange, Protein
Column Type Size-exclusion, Polyacrylamide Resin
MWCO 7.0 kDa
Product Line Pierce
Quantity 25 Columns
Sample Volume (Metric) 30 to 120 μL
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.