Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Pierce™ Premium Grade TCEP-HCl

Catalog No. PIPG82089 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 g
10 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPG82089 10 g
PIPG82080 1 g
PIPG82090 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PIPG82089 Supplier Thermo Scientific™ Supplier No. PG82089
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Optimize applications where product integrity and risk minimization are paramount with our highest quality formulation of this disulfide reducing agent.

Thermo Scientific Pierce Premium Grade TCEP-HCl is our highest quality formulation of this disulfide reducing agent, specially characterized for applications where product integrity and risk minimization are paramount.

Features of Premium Grade TCEP-HCl:

  • TCEP—a thiol-free, odorless yet potent reducing agent for cleaving disulfide bonds
  • High quality—identity and purity confirmed by several tests, including quantitative NMR
  • Product integrity—enhanced level of testing and characterization compared to standard grade
  • Lot retention—ample supply of past lots retained to ensure future process testing
  • Change management—Change Control Notification (CCN) service
  • Consistent manufacture—batch-specific manufacturing documentation review

Compared to the standard grade product, Premium Grade TCEP-HCl provides more clearly defined quality and product support by including (a) increased analytical testing and product characterization, (b) greater batch-specific information and quality assurance review, (c) extensive lot sample retention and (d) change control notification. TCEP-HCl is pure, crystalline Tris(2-carboxyethyl)phosphine hydrochloride, a thiol-free compound that is highly effective at reducing protein and peptide disulfide bonds, as well as being odorless and stable. TCEP selectively and completely reduces even the most stable water-soluble alkyl disulfides over a wide pH range.

Advantages of TCEP-HCl over other reducing agents:

  • Odorless—Unlike DTT or BME, TCEP is odor-free, so reductions can be carried out conveniently on the bench top.
  • Stable in air—The inherent stability of the TCEP moiety eliminates the need for any special precautions to avoid oxidation when handling, using or storing TCEP.
  • Efficient—For most applications, 5 to 50 mM TCEP provides sufficient molar excess to effectively reduce peptide or protein disulfide bonds within a few minutes at room temperature.
  • Compatible—With TCEP, removal of the reducing agent is not necessary prior to most applications, (e.g. histidine-tagged protein purification, maleimide conjugations).

Related Products

  • Pierce™ TCEP-HCl (Cat. No. 20490)
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Upon receipt store product at room temperature in sealed container to prevent oxidation. Product is shipped at ambient temperature.
Product Line Pierce
Form Solid
Quantity 10 g
Product Type Disulfide Reducing Reagent
Can other reducing agents other than DTT or BME be used to reduce proteins prior to electrophoresis? For example, what about TCEP (Tris Carboxy Ethyl Phosphene)?

TCEP, Tris Carboxy Ethyl Phosphene is an alternative sulfhydryl reducing agent for protein samples. It is an extremely potent and effective reducing agent for particularly ‘difficult' proteins. It is compatible with the Tris-Glycine gels and NuPAGE gels. It should be added to the sample buffer for these systems. 20 mM final (maximum) concentration is sufficient for samples. You may add alkylating agents, e.g. Iodine (50 mM Iodoacetic acid), to prevent re-forming of S-S bonds but it is not necessary. Do not heat because this will hydrolyze much of your sample. Instead let the sample sit for several minutes at RT and then load.

WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.