Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Pierce™ Protease Inhibitor XL Capsules, EDTA-free

Catalog No. PIA37989 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 Capsules
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIA37989 10 Capsules
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIA37989 Supplier Thermo Scientific™ Supplier No. A37989
Get 16% Off Protein Research Essentials
Use promo code PROTEIN16
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Includes

10 Capsules

Pierce Protease Inhibitor XL Capsules protect proteins from proteolytic degradation through inhibition of the serine, cysteine, and aspartic acid proteases, and aminopeptidases that are typically present in cell lysate samples. Each tablet is sufficient for 500 mL of solution

  • Broad spectrum formulation—contains AEBSF, bestatin, E-64 and pepstatin A for better protection versus PMSF
  • Large volume format – one capsule is sufficient for 500mL of solution
  • Convenient—ready-to-use, dissolves clearly into solution within 3 minutes
  • Compatible—use directly with Pierce Protein Assays
  • EDTA-free—formulated without EDTA, a metalloproteinase inhibitor 
TRUSTED_SUSTAINABILITY

Specifications

Product Type Protease Inhibitor Cocktail
Content And Storage 1 Vial, Store at 2°C to 8°C
Shipping Condition Room Temperature
Form Solid
Product Line Pierce
Sufficient For 500 mL Solution
Inhibitors Protease Inhibitor
Quantity 10 Capsules

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.