Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Pierce Protease and Phosphatase Inhibitor Mini Tablets

Catalog No. PIA32959 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
20 tablets
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIA32959 20 tablets
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
For Research Use Only. Not for use in diagnostic procedures.
Catalog No. PIA32959 Supplier Thermo Scientific™ Supplier No. A32959
Get 16% Off Protein Research Essentials
Use promo code PROTEIN16
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

For Research Use Only. Not for use in diagnostic procedures.

Thermo Scientific™ Pierce Protease and Phosphatase Inhibitor Mini Tablets broad-spectrum protease and pghospahatase inhibitor formulation provides complete protection from dephosphorylation and proteolytic degradation.

The Pierce Protease and Phosphatase Inhibitor Mini Tablets contain aprotinin, bestatin, E-64, EDTA, and leupeptin to provide protection from degradation by serine-proteases, cysteine-proteases, aspartic acid-proteases. To prevent the dephosphorylation activity of serine/threonine and tyrosine phosphatases, each tablet also contains sodium fluoride, sodium orthovanadate, sodium pyrophosphate and beta-glycerophosphate. Pierce Protease and Phosphatase Inhibitor Tablets are stable at 4°C for up to 12 months, and each tablet is sufficient for 10mL of solution.

Highlights:
  • Improved formulation&mdsashtablets dissolve quickly into a clear solution
  • Economical – more cost-effective than other commercially available formulations for equivalent volumes
  • Convenient&mdsashready-to-use, fully disclosed, broad-spectrum formulations
  • Compatible&mdsashuse directly with Thermo Scientific™ Pierce™ BCA Assays
* EDTA is present only in Product No. A32959.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Protease and Phosphatase Inhibitor
Content And Storage 1 Vial, Store at 2°C to 8°C
Shipping Condition Wet Ice
Form Solid
Product Line Pierce
Sufficient For 10 mL Solution
Inhibitors Protease Inhibitor, Phosphatase Inhibitor
Quantity 20 tablets

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.