Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Pierce™ Protein Concentrator PES

Catalog No. PI88535S Shop All Thermo Scientific Products
Change view
Click to view available options
MWCO:
10 kDa
5 kDa
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. MWCO
PI88535S 10 kDa
PI88534S 5 kDa
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PI88535S Supplier Thermo Scientific™ Supplier No. 88535S
Get 16% Off Protein Research Essentials
Use promo code PROTEIN16
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Thermo Scientific™ Pierce™ Protein Concentrators PES are easy-to-use centrifugal devices that provide fast processing and excellent recovery of protein samples.

The Thermo Scientific™ Pierce™ concentrators are easy-to-use centrifugal devices that provide fast processing and excellent recovery of protein samples. These disposable ultrafiltration devices contain a polyethersulfone (PES) membrane for the concentration, desalting, and buffer exchange of biological samples, such as tissue culture media, antiserum, monoclonal antibody preparations, and chromatography fractions. They can also be used to remove unincorporated label following protein modification reactions. 

The PES membrane in these devices is available in five distinct molecular-weight cutoffs (MWCO) of 3K, 5K, 10K, 30K, and 100K, and can be used for processing sample volumes from 100 μL to 100 mL. The MWCOs are etched on the sides of the concentrators for easy identification, while a clear window with graduations on the side of each device allows for visual determination of the concentrated sample (retentate) volume. The unique design provides reliable and consistent results. Multiple device sizes are available to handle maximum sample volumes of 0.5 mL, 6 mL, 20 mL, and 100 mL.
  • Rapid processing—unique design minimizes membrane fouling and sample concentration of 10- to 30-fold can be achieved in 5–30 minutes for 10K MWCO (device dependent—times may vary for other MWCO’s), even with particle laden solutions.
  • High recovery—retain >90% of protein samples while removing contaminants or exchanging buffers.
  • Convenient—clear markings, wide sample chamber and removable filtrate chamber make handling simple and easy.
  • Instrument compatible—can be used with standard centrifuges utilizing either fixed-angle or swinging-bucket rotors.
TRUSTED_SUSTAINABILITY

Specifications

Material (Membrane) PES
MWCO 10 kDa
For Use With (Application) concentration, desalting, and buffer exchange of biological samples
Quantity 1pk
Type Spin Column, Bottle
Volume (Metric) 20 to 100 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.