Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Pierce™ Protein L Magnetic Beads
GSA_VA

Catalog No. PI88850 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI88850 5 mL
PI88849 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PI88850 Supplier Thermo Scientific™ Supplier No. 88850
Get 15% Off Sample Preparation Products
Use promo code SAMPREP15
View all promotions
Add to Cart
Edge
Add to Cart

Perform antibody purification using manual or robotic magnetic separators with these high-capacity and high-throughput affinity particles.

Thermo Scientific Pierce Protein L Magnetic Beads are high-capacity and high-throughput affinity particles for antibody purification using manual or robotic magnetic separators.

Features of Protein L Magnetic Beads:

  • Selective—immobilized Protein L is ideal for selective purification of human and mouse antibodies that have kappa light chains
  • Low non-specific binding—stable, pre-blocked beads provide clean purification of antibody
  • Versatile—beads are compatible with manual and automated workflows (e.g., Thermo Scientific KingFisher Instruments)

Recombinant Protein L, covalently attached to a blocked magnetic bead surface, selectively binds mouse and human antibodies through kappa light chains. The beads are commonly used to purify monoclonal antibodies in cell culture supernatants supplemented with bovine serum as Protein L does not bind bovine IgG. The Pierce Protein L Magnetic Beads can be used manually with a magnetic stand, as well as with automated platforms such as the Thermo Scientific KingFisher Instruments for high-throughput workflows.

Applications:

  • Purification of monoclonal and polyclonal antibodies that bind poorly to Protein A or Protein G magnetic beads
  • Purification of monoclonal antibodies from culture supernatants supplemented with bovine serum
  • Protein L IP and co-IP assays
  • Purification of ScFv and Fab fragments containing kappa light chains

Thermo Scientific Pierce Protein L Magnetic Beads are typically used for purifying mouse and human antibodies containing kappa light chains from serum, cell culture supernatant or ascites. Protein L can bind a broader range of Ig classes than Protein A or Protein G, including IgG, IgM, IgA, IgE and IgD. Protein L binds strongly to human (kappa I, III and IV only), mouse (kappa I only), rat and pig immunoglobulins. It binds weakly to rabbit immunoglobulins and does not bind bovine, goat or sheep immunoglobulins. Single chain variable fragments (scFv) and Fab fragments also bind to Protein L. The protocol for the Pierce Protein L beads has been optimized to allow for high recovery and high purity of isolated antibodies.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 5 mL
Concentration Slurry: 10mg/mL, 1% solids
Content And Storage Upon receipt store at 4°C. Product shipped with an ice pack.
Particle Size 1 μm
Form Liquid
Product Line Pierce
Quantity 5 mL
Protein Form Recombinant
Target Antibody
Type Magnetic Affinity Bead
Ligand Type Protein L
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.