Learn More
Thermo Scientific™ Sodium Acetate (3 M)
Catalog No. PI17888 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 L
Catalog No. Quantity
PI17888 1 L
1 options
Catalog No. PI17888 Supplier Thermo Scientific™ Supplier No. 17888
Consitently purify and store DNA without any worries. Thermo Scientific™ Pierce™ Sodium Acetate Solution (3M) is a highly purified, sterile 3M sodium acetate solution.
Thermo Scientific™ Pierce Sodium Acetate Solution is a sterile, 3M soluion of sodium acetate that is useful for DNA purification and storage.
Additional Information and Online Ordering
Thermo Scientific™ Pierce™ Sodium Acetate Solution is a sterile, 3M solution of sodium acetate that is useful for DNA purification and storage. The clear, colorless and odorless liquid is delivered in 1L bottles.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 20 to 25°C.
Product Type Sodium Acetate Solution
Quantity 1 L
Form Liquid