Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Sodium Dodecyl Sulfate (SDS), C12

Catalog No. PI28312 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI28312 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI28312 Supplier Thermo Scientific™ Supplier No. 28312
Get 16% Off Protein Research Essentials
Use promo code PROTEIN16
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Highly purified form of SDS especially suited for protein experiments requiring tightly controlled solubility parameters.

Thermo Scientific™ Sodium Dodecyl Sulfate (C12) is a highly purified form of SDS that is especially suited for protein experiments requiring tightly controlled solubility parameters.

Sodium dodecyl sulfate (SDS) is an alkyl sulfate, anionic detergent used frequently in protein electrophoresis and protein solubilization methods. It is a component of many cell lysis buffer formulations, including RIPA buffer. Typical SDS or lauryl sulfate preparations are mixture of several different chain lengths (C10 to C18). However, protein renaturation and other specialized methods require very specific surfactant and buffer conditions. This particular formulation of SDS is highly purified to eliminate nearly all secondary chain lengths besides C12.

Highlights:

  • Popular anionic detergent for protein electrophoresis and solubilization
  • Component of many cell lysis buffer formulations
  • Specially purified formulation is >98% C12 alkyl sulfate
  • Contains low levels of hexadecyl sulfate (C16), which inhibits protein renaturation

Detergent properties (pure C12):

  • Molecular Weight: 288.5
  • Detergent Class: Ionic (anionic)
  • Aggregation Number: 62
  • Micelle Molecular Weight: 18,000
  • Critical Micelle Concentration (CMC): 6 to 8mM (0.1728 to 0.2304%, w/v)
  • Cloud Point: >100°C
  • Dialyzable: No
TRUSTED_SUSTAINABILITY

Specifications

Product Type Detergent
Content And Storage Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 20°C to 25°C.
Form Powder
Reagent Type Detergent (Pure)
Quantity 500 g

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.