Learn More
Description
Streptavidin Protein, AP conjugate, 21324, from Invitrogen., 21324, from Invitrogen. Species Reactivity: ; Applications: ELISA, Western Blot.
Formulation: 1mg purified protein (2-4mg/mL) in TBS with 50% glycerol, 0.1mMmgCl2, 0.1mM ZnCl2 and no preservative; pH 8
This product has been successfully used in ELISA and Western Blot procedures.
Specifications
Specifications
| Concentration | 0.1 μg/mL |
| Conjugate | AP |
| Quantity | 1 mg |
| Content And Storage | Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, but do not store above -20°C. |
| Form | Liquid |
| Product Line | Pierce |
| Product Type | Streptavidin Conjugate |
| Target Molecule | Biotin |
Safety and Handling
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.