Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Pierce™ Sulfhydryl Addition Kit

Catalog No. PI23460 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 Reaction
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI23460 1 Reaction
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI23460 Supplier Thermo Scientific™ Supplier No. 23460
Season of Savings
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Modify protein primary amines to create free sulfhydryl groups for labeling, crosslinking or immobilization techniques; complete kit with SATA reagent and hydroxylamine.

Thermo Scientific™ Pierce Sulfhydryl Addition Kit is a complete set of reagents, including SATA, to enable modification of primary amines to free sulfhydryl groups for use in heterobifunctional crosslinking techniques.

Highlights:

  • Reacts with primary amines to add protected sulfhydryls
  • An easy deprotection step generates a free sulfhydryl group
  • Thermo Scientific™ Sulfhydryl Addition Kit contains all necessary components for thiolation, deprotection, purification and quantitation
TRUSTED_SUSTAINABILITY

Specifications

Chemical Reactivity Amine
Content And Storage Sufficient For: One modification experiment with 0.5 to 5 mg of purified protein
• SATA, 2 mg
• Hydroxylamine·HCl, 5 mg
• Conjugation Buffer Stock (10X), 20 mL
• PBS pack (makes 500 mL), 1 pack
• Dimethylformamide (DMF), 1 mL
• Dextran Desalting Column, 1 x 5 mL
• Column Extender, 1
• Ellman's Reagent (DTNB), 2 mg
• Cysteine·HCl, 20 mg

Store at 4°C.
Label or Dye Sulfhydryl Group
Label Type Conjugating Groups
Product Line Pierce
Crosslinker Type Heterobifunctional
Reactive Moiety Active Ester, Succinimidyl Ester, NHS Ester
Spacer Short (<10 Å)
Form Powder
Reagent Type SATA
Quantity 1 Reaction
Solubility DMF, DMSO
Product Type Crosslinker
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.