Learn More
Description
Thermo Scientific™ Pierce Vacuum Hydrolysis Tubes are high-quality glass tubes designed for heated, acid-based, vacuum hydrolysis of proteins and peptides for amino acid analysis.
Highlights:
- Upper temperature limit of the Vacuum Hydrolysis Tubes is 260°C; however, do not heat the tubes greater than 110°C in an oven Vacuum hydrolysis tubes fit conveniently into Reacti-Block™ Aluminum Heating Blocks
Specifications
Specifications
| Description | Vacuum Hydrolysis Tube, 18mL |
| Product Type | Vacuum Hydrolysis Tube |
| Content And Storage | 0.0 |
| Format | Tube |
| Quantity | 1 Tube |
Safety and Handling
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.