Learn More
Thomson Instrument Company Pierceable Foil Heat Seal | Use w/ All Plates
SDP
Catalog No. NC3059856
Encompass_Preferred
Thomson Instrument Company Foil Heat Seals are for short and long-term storage in well plates. Thomson well plates are offered in 24 and 96-well configurations with a V, U or pyramid shape bottom, and square or round wells for cell culture and analytical sample preparation. Well plates are designed to SLAS specifications and work well on automated liquid handling systems. Many plates are individually wrapped for your convince, and ideally designed to work with Thomson filter plates. Thomson manufactures its products in the USA to assure same day shipping.
Catalog No. NC3059856 Supplier Thomson Instrument Company Supplier No. 899403
Thomson Instrument Company Foil Heat Seals are for short and long-term storage in well plates. Thomson well plates are offered in 24 and 96-well configurations with a V, U or pyramid shape bottom, and square or round wells for cell culture and analytical sample preparation. Well plates are designed to SLAS specifications and work well on automated liquid handling systems. Many plates are individually wrapped for your convince, and ideally designed to work with Thomson filter plates. Thomson manufactures its products in the USA to assure same day shipping.