Learn More
Sigma Aldrich Fine Chemicals Biosciences PierceASeal foilTM heat se
Catalog No. 502901278
Encompass
Pierceable foil seal with good solvent resistance.features foil high solvent resistance including DMSO non-peelable multiple sealing/re-sealing permissible easy sealing surface identification pierceable.Compatibility Polypropylene (PP) Polystyrene (PS). Seal integrity range -20C to 120C.Application PCR compound storage sample shipping.
Catalog No. 50-290-1278 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z740334100EA
Pierceable foil seal with good solvent resistance.features foil high solvent resistance including DMSO non-peelable multiple sealing/re-sealing permissible easy sealing surface identification pierceable.Compatibility Polypropylene (PP) Polystyrene (PS). Seal integrity range -20C to 120C.Application PCR compound storage sample shipping.