Learn More
Perkin Elmer US LLC Pierced Aluminum Covers - Qty. 400
Catalog No. 502111767
Encompass_Preferred
Most vented or pierced pans and covers use the Universal Crimper.
Catalog No. 50-211-1767 Supplier Perkin Elmer US LLC Supplier No. B7001014
Most vented or pierced pans and covers use the Universal Crimper.