Learn More
Sklar Instruments Vasectomy Piercing Forceps, Sklar, 5-1/2 in., Sharp, Curved, Smooth, Stainless Steel. EA/1.
SDP
Catalog No. 501436506 Shop All Sklar Instruments Products
Encompass_Preferred
Vasectomy Piercing Forceps, Sklar, 5-1/2 in., Sharp, Curved, Smooth, Stainless Steel. Sklar's Vasectomy Piercing Forceps are a ratcheted, finger ring instrument with sharp tips used to pierce the scrotal skin in sutureless vasectomy procedures. It is used in conjunction with the vasectomy clamp with ring tips. This product is curved with sharp tips and 5-1/2 inches long.
This item is not returnable. View return policy
Catalog No. 50-143-6506 Supplier Sklar Instruments Supplier No. 856224
Vasectomy Piercing Forceps, Sklar, 5-1/2 in., Sharp, Curved, Smooth, Stainless Steel. Sklar's Vasectomy Piercing Forceps are a ratcheted, finger ring instrument with sharp tips used to pierce the scrotal skin in sutureless vasectomy procedures. It is used in conjunction with the vasectomy clamp with ring tips. This product is curved with sharp tips and 5-1/2 inches long.