Learn More
Enzo Life Sciences Piericidin A (2mg). CAS: 2738-64-9
SDP
Catalog No. 502008984
Encompass_Preferred
Antibiotic. Specific and potent inhibitor of NADH-ubiquinone oxidoreductase (complex I) that binds to ubiquinone binding site(s). Inhibits both mitochondrial and bacterial NADH ubiquinone oxidoreductase. Anticancer agent that prevents up-regulation of GRP78, and exhibits cytotoxic activity for etoposide resistant cancer cells under glucose-deprived conditions. Alternative name: Piericidin A1, Shaoguanmycin B, Antibiotic MT 1882-I, Antibiotic SN 198E, Antibiotic IT 143D. Purity: ≥95% (HPLC). Appearance: Pale yellow oil. Solubility: Soluble in 100% ethanol, methanol, dimethyl formamide or DMSO. Poorly soluble in water. Long Term Storage: -20°C.
Catalog No. 50-200-8984 Supplier Enzo Life Sciences Supplier No. ALX380235M002
Add to Cart
Add to Cart
Antibiotic. Specific and potent inhibitor of NADH-ubiquinone oxidoreductase (complex I) that binds to ubiquinone binding site(s). Inhibits both mitochondrial and bacterial NADH ubiquinone oxidoreductase. Anticancer agent that prevents up-regulation of GRP78, and exhibits cytotoxic activity for etoposide resistant cancer cells under glucose-deprived conditions. Alternative name: Piericidin A1, Shaoguanmycin B, Antibiotic MT 1882-I, Antibiotic SN 198E, Antibiotic IT 143D. Purity: ≥95% (HPLC). Appearance: Pale yellow oil. Solubility: Soluble in 100% ethanol, methanol, dimethyl formamide or DMSO. Poorly soluble in water. Long Term Storage: -20°C.