Learn More
ATCC Piezo1 knock-out HEK293T; Embryonic; Kidney; Human (Homo sapiens)
Catalog No. 502384764
Encompass
The Piezo1 knock-out HEK293T cell line can be used to assess ion channel activity by electrophysiology without any interference from endogenous mechanically activated currents caused by the ion channel Piezo1.
Catalog No. 50-238-4764 Supplier ATCC Supplier No. CRL3519
The Piezo1 knock-out HEK293T cell line can be used to assess ion channel activity by electrophysiology without any interference from endogenous mechanically activated currents caused by the ion channel Piezo1.