Learn More
Cayman Chemical PIfIthrIn-a 10mg
SDP
Catalog No. 502952323
Encompass_Preferred
An inactivator of p53 that blocks p53-dependent transcriptional activation and apoptosis, preventing p53-mediated apoptosis by cytotoxic compounds in C8 cells at 10 μM and in human umbilical vein endothelial cells at 30 μM
Catalog No. 50-295-2323 Supplier Cayman Chemical Supplier No. 1332610 MG
An inactivator of p53 that blocks p53-dependent transcriptional activation and apoptosis, preventing p53-mediated apoptosis by cytotoxic compounds in C8 cells at 10 μM and in human umbilical vein endothelial cells at 30 μM