Learn More
Selleck Chemical LLC Pifithrin-a (PFTa) HBr S2929-1g
SDP
Catalog No. 503111625
Encompass_Preferred
Pifithrin- is an inhibitor of p53 inhibiting p53-dependent transactivation of p53-responsive genes Pifithrin- is also a potent agonist of the aryl hydrocarbon receptor (AhR)
Catalog No. 50-311-1625 Supplier Selleck Chemical LLC Supplier No. S29291G
Pifithrin- is an inhibitor of p53 inhibiting p53-dependent transactivation of p53-responsive genes Pifithrin- is also a potent agonist of the aryl hydrocarbon receptor (AhR)