Learn More
Enzo Life Sciences Pifithrin-alpha (cyclic). hydrobromide (10mg). CAS: 60477-34-1
SDP
Catalog No. 502008466
Encompass_Preferred
Stable analog of pifithrin-α (BML-GR325), with biological properties similar to its parent compound, but with reduced cytotoxicity. Inhibitor of p53. Reversibly inhibits p53-dependent transactivation of p53-responsive genes and reversibly blocks p53-mediated apoptosis. Potent STAT6 transcriptional inhibitor. Increases apoptosis in target cells when used in combination with antimicrotubule agents. Formula: C16H16N2S . HBr. MW: 268.4 . 80.9. Purity: ≥95% (HPLC, TLC). Appearance: White to off-white powder. Solubility: Soluble in DMSO (100mg/ml). Long Term Storage: -20°C. Handling: Protect from light and moisture. Keep under inert gas.
Catalog No. 50-200-8466 Supplier Enzo Life Sciences Supplier No. ALX270287M010
Stable analog of pifithrin-α (BML-GR325), with biological properties similar to its parent compound, but with reduced cytotoxicity. Inhibitor of p53. Reversibly inhibits p53-dependent transactivation of p53-responsive genes and reversibly blocks p53-mediated apoptosis. Potent STAT6 transcriptional inhibitor. Increases apoptosis in target cells when used in combination with antimicrotubule agents. Formula: C16H16N2S . HBr. MW: 268.4 . 80.9. Purity: ≥95% (HPLC, TLC). Appearance: White to off-white powder. Solubility: Soluble in DMSO (100mg/ml). Long Term Storage: -20°C. Handling: Protect from light and moisture. Keep under inert gas.