Learn More
Apexbio Technology LLC Pifithrin-α (PFTα) 63208-82-2 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062633
Encompass_Preferred
Pifithrin- (PFT CAS 63208-82-2) is a synthetic water-soluble inhibitor targeting the tumor suppressor protein p53 Initially identified by its ability to block activation of p53-responsive genes PFT reduces apoptosis in transformed mouse embryonic fibroblasts (expressing E1a and ras) in a p53-dependent manner Additionally PFT prevents DNA damage-induced growth arrest in human diploid fibroblasts and promotes G2-phase cell cycle arrest following -irradiation It also decreases the expression of pluripotency marker Nanog affecting self-renewal potential in embryonic stem cells without compromising viability Thus PFT serves as a valuable research tool to investigate p53-mediated cellular pathways
Catalog No. 50-000-62633 Supplier Apexbio Technology LLC Supplier No. A42065.1
Pifithrin- (PFT CAS 63208-82-2) is a synthetic water-soluble inhibitor targeting the tumor suppressor protein p53 Initially identified by its ability to block activation of p53-responsive genes PFT reduces apoptosis in transformed mouse embryonic fibroblasts (expressing E1a and ras) in a p53-dependent manner Additionally PFT prevents DNA damage-induced growth arrest in human diploid fibroblasts and promotes G2-phase cell cycle arrest following -irradiation It also decreases the expression of pluripotency marker Nanog affecting self-renewal potential in embryonic stem cells without compromising viability Thus PFT serves as a valuable research tool to investigate p53-mediated cellular pathways