Learn More
Enzo Life Sciences Pifithrin-alpha (25mg). CAS: 63208-82-2
SDP
Supplier:  Enzo Life Sciences BMLGR3250025
Encompass_Preferred
Chemical inhibitor of p53 that protects mice from the side effects of cancer therapy. Reversibly blocks p53-dependent transcriptional activation and apoptosis. Protects against neuronal death in models of stroke and neurodegenerative disorders. Suppresses self-renewal of embryonic stem cells. Alternative name: 1-(4-Methylphenyl)-2-(4,5,6,7-tetrahydro-2-imino-3(2H)-benzothiazolyl)ethanone . HBr, PFT-alpha. Purity: ≥98% (TLC). Solubility: Soluble in DMSO (>25mg/ml) or methanol. Long Term Storage: -20°C.
Catalog No. 50-201-0979