Learn More
AdipoGen Pifithrin-alpha cyclic HBr
SDP
Catalog No. 502562374
Encompass_Preferred
Chemical. CAS 511296-88-160477-34-1 free base. Formula C16H16N2S . HBr. MW 268.4 . 80.9. Stable analog of pifithrin-alpha. p53 inhibitor.
Catalog No. 50-256-2374 Supplier AdipoGen Supplier No. AGCR10052M010
Chemical. CAS 511296-88-160477-34-1 free base. Formula C16H16N2S . HBr. MW 268.4 . 80.9. Stable analog of pifithrin-alpha. p53 inhibitor.