Learn More
AdipoGen Pifithrin-alpha HBr
SDP
Catalog No. 502562401
Encompass_Preferred
Chemical. CAS 63208-82-2. Formula C16H18N2OS . HBr. MW 286.4 . 80.9. p53 inhibitor. Reversibly blocks p53-dependent transcriptional activation. Tool to combat side effects of cancer therapy. Apoptosis inhibitor.
Catalog No. 50-256-2401 Supplier AdipoGen Supplier No. AGCR10004M025
Chemical. CAS 63208-82-2. Formula C16H18N2OS . HBr. MW 286.4 . 80.9. p53 inhibitor. Reversibly blocks p53-dependent transcriptional activation. Tool to combat side effects of cancer therapy. Apoptosis inhibitor.