Learn More
Abcam Pifithrin-alpha-HBr, p53 Inhibitor, 10MG
Catalog No. 502707564
Encompass_Preferred
MW 367.3 Da, Purity >95%. p53 Inhibitor. Specifically blocks tumor supressor p53 transcriptional activity and can supress heat shock and glucocorticoid signalling pathways. Additionally offers protection to cells from p53 mediated apoptosis. Active in vivo; protects mice from chemotherapy side effects associated with p53 induction. Also aryl hydrocarbon receptor (AHR) agonist.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7564 Supplier Abcam Supplier No. AB12047810MG
MW 367.3 Da, Purity >95%. p53 Inhibitor. Specifically blocks tumor supressor p53 transcriptional activity and can supress heat shock and glucocorticoid signalling pathways. Additionally offers protection to cells from p53 mediated apoptosis. Active in vivo; protects mice from chemotherapy side effects associated with p53 induction. Also aryl hydrocarbon receptor (AHR) agonist.
The product is subject to the following: Abcam Restricted Use Statement