Learn More
TARGETMOL CHEMICALS INC PIFITHRIN-ALPHA HYDROBRO 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T270725MG
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Pifithrin-(alpha) hydrobromide (Pifithrin-(alpha) hydrobromide) is a p53 inhibitor inhibiting p53-dependent transactivation of p53-responsive genes. purity: 98%
Catalog No. 50-258-0651