Learn More
TARGETMOL CHEMICALS INC Research Chemical 10mg
SDP
Supplier:  TARGETMOL CHEMICALS INC T1247210MG
Encompass_Preferred
Also available in 1 mg, 2 mg, 5 mg, 25 mg, 50 mg, 100 mg, 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Pifithrin-(alpha), p-Nitro, Cyclic (PFN-(alpha)) (PFN-(alpha)) is cell-permeable and active-form inhibitor of p53. purity: 99%
Catalog No. 50-303-8153