Learn More
Medchemexpress LLC Pifithrin-β hydrobromide | 511296-88-1 | 100.0% | 349.29 | 100 MG
SDP
Catalog No. 5000390943
Encompass_Preferred
Pifithrin-β hydrobromide is a potent p53 inhibitor with an IC50 of 23 μM. It is considered a lead compound for cancer and neurodegenerative disease therapy. This compound, which rapidly converts from its unstable alpha form, has demonstrated neuroprotective effects in viability assays.
  • Potent p53 inhibitor
  • IC50 of 23 μM
  • Lead compound for cancer and neurodegenerative disease research
  • Exerts neuroprotective effects
Catalog No. 50-003-90943 Supplier Medchemexpress LLC Supplier No. HY16702A100MG
Pifithrin-β hydrobromide is a potent p53 inhibitor with an IC50 of 23 μM. It is considered a lead compound for cancer and neurodegenerative disease therapy. This compound, which rapidly converts from its unstable alpha form, has demonstrated neuroprotective effects in viability assays.
  • Potent p53 inhibitor
  • IC50 of 23 μM
  • Lead compound for cancer and neurodegenerative disease research
  • Exerts neuroprotective effects