Learn More
Medchemexpress LLC Pifithrin-β hydrobromide | 511296-88-1 | 100.0% | 349.29 | 50 MG
SDP
Catalog No. 5000390797
Encompass_Preferred
Pifithrin-β hydrobromide, also known as PFT β hydrobromide, is a potent p53 inhibitor with an IC50 of 23 μM. It is related to Pifithrin-α, a lead compound for cancer and neurodegenerative disease therapy, which rapidly converts to Pifithrin-β, its N-acetyl derivative. Pretreatment with 1 and 10 μM pifithrin-β has shown neuroprotective effects.
  • Potent p53 inhibitor
  • Exerts neuroprotective effects
  • Used in cancer and neurodegenerative disease research
Catalog No. 50-003-90797 Supplier Medchemexpress LLC Supplier No. HY16702A50MG
Pifithrin-β hydrobromide, also known as PFT β hydrobromide, is a potent p53 inhibitor with an IC50 of 23 μM. It is related to Pifithrin-α, a lead compound for cancer and neurodegenerative disease therapy, which rapidly converts to Pifithrin-β, its N-acetyl derivative. Pretreatment with 1 and 10 μM pifithrin-β has shown neuroprotective effects.
  • Potent p53 inhibitor
  • Exerts neuroprotective effects
  • Used in cancer and neurodegenerative disease research