Learn More
Medchemexpress LLC Pifithrin-β hydrobromide | 511296-88-1 | 99.9% | 349.29 | 1 ML
SDP
Catalog No. 5000390696
Encompass_Preferred
Pifithrin-β hydrobromide is a potent p53 inhibitor with an IC50 of 23 μM. It is regarded as a lead compound for cancer and neurodegenerative disease therapy. Pifithrin-α, an inhibitor of the p53 protein, is unstable in culture medium and quickly converts to pifithrin-β. Studies show that pretreatments with 1 and 10 μM pifithrin-β can exert neuroprotective effects after 24 hours.
  • Potent p53 inhibitor with an IC50 of 23 μM.
  • Regarded as a lead compound for cancer and neurodegenerative disease therapy.
  • Exerts neuroprotective effects.
Catalog No. 50-003-90696 Supplier Medchemexpress LLC Supplier No. HY16702A10MM/1ML
Pifithrin-β hydrobromide is a potent p53 inhibitor with an IC50 of 23 μM. It is regarded as a lead compound for cancer and neurodegenerative disease therapy. Pifithrin-α, an inhibitor of the p53 protein, is unstable in culture medium and quickly converts to pifithrin-β. Studies show that pretreatments with 1 and 10 μM pifithrin-β can exert neuroprotective effects after 24 hours.
  • Potent p53 inhibitor with an IC50 of 23 μM.
  • Regarded as a lead compound for cancer and neurodegenerative disease therapy.
  • Exerts neuroprotective effects.