Learn More
Medchemexpress LLC Pifithrin-β hydrobromide | 511296-88-1 | MFCD02683960 | >=98.0% | 349.29 g/mol | C16H17BrN2S | 10 MG
SDP
Catalog No. 5000288949
Encompass_Preferred
Pifithrin-β hydrobromide is a small-molecule inhibitor of the p53 pathway supplied as the hydrobromide salt. It reversibly blocks p53 transcriptional activity (reported IC50 = 23 μM) and is used as a research tool in cellular and biochemical studies of apoptosis and ferroptosis. Chemical formula C16H17BrN2S; molecular weight 349.29 g/mol; CAS 511296-88-1.
  • Reversible p53 inhibitor with reported IC50 of 23 μM.
  • Hydrobromide salt for improved solubility and handling.
  • Reported high purity (≥98% by HPLC) in supplier listings.
  • Suitable for cellular and biochemical assays studying apoptosis and ferroptosis.
  • Store sealed at 4 °C as a solid; in solution store at -80 °C for long-term stability.
Catalog No. 50-002-88949 Supplier Medchemexpress LLC Supplier No. HY16702A10MG
Pifithrin-β hydrobromide is a small-molecule inhibitor of the p53 pathway supplied as the hydrobromide salt. It reversibly blocks p53 transcriptional activity (reported IC50 = 23 μM) and is used as a research tool in cellular and biochemical studies of apoptosis and ferroptosis. Chemical formula C16H17BrN2S; molecular weight 349.29 g/mol; CAS 511296-88-1.
  • Reversible p53 inhibitor with reported IC50 of 23 μM.
  • Hydrobromide salt for improved solubility and handling.
  • Reported high purity (≥98% by HPLC) in supplier listings.
  • Suitable for cellular and biochemical assays studying apoptosis and ferroptosis.
  • Store sealed at 4 °C as a solid; in solution store at -80 °C for long-term stability.