Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pifithrin-α hydrobromide | 63208-82-2 | C16H19BrN2OS | 1 G
SDP

Catalog No. 5000376484
Encompass_Preferred

Pifithrin-α hydrobromide is a chemical compound known for its role as a p53 inhibitor, effectively blocking its transcriptional activity and preventing apoptosis in cells. Additionally, it functions as an aryl hydrocarbon receptor (AhR) agonist.

  • Purity of 98.58%
  • Appears as a solid, white to light yellow in color
  • Recommended storage at 4°C, sealed and away from moisture; in solvent at -80°C for 6 months or -20°C for 1 month
  • Soluble in DMSO at concentrations ≥ 50 mg/mL
  • Targets p53 and aryl hydrocarbon receptor (AhR)
  • Suppresses p53 protein transcription and inhibits p53-dependent apoptosis
  • Acts as a potent AhR agonist, inducing DNA binding complex formation

Catalog No. 50-003-76484 Supplier Medchemexpress LLC Supplier No. HY154841G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pifithrin-α hydrobromide is a chemical compound known for its role as a p53 inhibitor, effectively blocking its transcriptional activity and preventing apoptosis in cells. Additionally, it functions as an aryl hydrocarbon receptor (AhR) agonist.

  • Purity of 98.58%
  • Appears as a solid, white to light yellow in color
  • Recommended storage at 4°C, sealed and away from moisture; in solvent at -80°C for 6 months or -20°C for 1 month
  • Soluble in DMSO at concentrations ≥ 50 mg/mL
  • Targets p53 and aryl hydrocarbon receptor (AhR)
  • Suppresses p53 protein transcription and inhibits p53-dependent apoptosis
  • Acts as a potent AhR agonist, inducing DNA binding complex formation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.