Learn More
Medchemexpress LLC Pifithrin-α (hydrobromide) | 63208-82-2 | C16H19BrN2OS | 50 MG
SDP
Catalog No. 5000376601
Encompass_Preferred
Pifithrin-α hydrobromide is a p53 inhibitor that blocks its transcriptional activity and prevents cells from apoptosis. It also acts as an aryl hydrocarbon receptor (AhR) agonist.
  • Blocks p53 transcriptional activity.
  • Prevents cells from apoptosis.
  • Acts as an aryl hydrocarbon receptor (AhR) agonist.
  • Suppresses glucose oxidase (GOX)-induced p53 protein increase.
  • Reduces Bcl-2 protein and prevents Bax increase.
  • Up-regulates the classic AhR target gene CYP1A1.
  • Demonstrates in vivo efficacy in reducing aortic rupture and normalizing aortic morphology in Foxe3-/- mice.
Catalog No. 50-003-76601 Supplier Medchemexpress LLC Supplier No. HY1548450MG
Pifithrin-α hydrobromide is a p53 inhibitor that blocks its transcriptional activity and prevents cells from apoptosis. It also acts as an aryl hydrocarbon receptor (AhR) agonist.
  • Blocks p53 transcriptional activity.
  • Prevents cells from apoptosis.
  • Acts as an aryl hydrocarbon receptor (AhR) agonist.
  • Suppresses glucose oxidase (GOX)-induced p53 protein increase.
  • Reduces Bcl-2 protein and prevents Bax increase.
  • Up-regulates the classic AhR target gene CYP1A1.
  • Demonstrates in vivo efficacy in reducing aortic rupture and normalizing aortic morphology in Foxe3-/- mice.