Learn More
Medchemexpress LLC Pifithrin-α hydrobromide | 63208-82-2 | C16H19BrN2OS | 500 MG
SDP
Supplier:  Medchemexpress LLC HY15484500MG
Encompass_Preferred
Pifithrin-α hydrobromide is a p53 inhibitor that blocks its transcriptional activity, preventing cells from apoptosis. It also acts as an aryl hydrocarbon receptor (AhR) agonist. This water-soluble compound can suppress p53 protein transcription and glucose oxidase (GOX)-induced p53 protein increase.
  • Blocks p53 transcriptional activity
  • Prevents cells from apoptosis
  • Acts as an aryl hydrocarbon receptor (AhR) agonist
  • Suppresses p53 protein transcription
  • Blocks GOX-induced Bcl-2 protein reduction and prevents Bax increase
  • Binds to AhR and activates reporter activity
  • Up-regulates classic AhR target gene CYP1A1
Catalog No. 50-003-76197