Learn More
Selleck Chemical LLC Pifithrin-μ S2930-10mM/1mL
SDP
Catalog No. 503111826
Encompass_Preferred
Pifithrin- (NSC 303580 PFT 2-Phenylethynesulfonamide) is a specific p53 inhibitor by reducing its affinity to Bcl-xL and Bcl-2 and also inhibits HSP70 function and autophagy
Catalog No. 50-311-1826 Supplier Selleck Chemical LLC Supplier No. S293010MM/1ML
Pifithrin- (NSC 303580 PFT 2-Phenylethynesulfonamide) is a specific p53 inhibitor by reducing its affinity to Bcl-xL and Bcl-2 and also inhibits HSP70 function and autophagy