Learn More
Abcam Pifithrin-mu, Inhibitor of p53-mediated apoptosis, 10MG
Catalog No. 502707167
Encompass_Preferred
MW 181.21 g/mol, Purity >98%. An inhibitor of p53-mediated apoptosis. Prevents p53 binding at the mitochondrial surface without affecting p53 transactivational activities. Pifithrin-μ reduces p53-mediated apoptosis triggered by nutlin; also interacts selectively with HSP70, disrupting its association with many substrate proteins.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7167 Supplier Abcam Supplier No. AB12088610MG
MW 181.21 g/mol, Purity >98%. An inhibitor of p53-mediated apoptosis. Prevents p53 binding at the mitochondrial surface without affecting p53 transactivational activities. Pifithrin-μ reduces p53-mediated apoptosis triggered by nutlin; also interacts selectively with HSP70, disrupting its association with many substrate proteins.
The product is subject to the following: Abcam Restricted Use Statement