Learn More
Medchemexpress LLC HY-123076 5mg Medchemexpress, Pifithrin-α, p-Nitro, Cyclic CAS:60477-38-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1230765MG
Encompass_Preferred
Medchemexpress, HY-123076 5mg Pifithrin-α, p-Nitro, Cyclic CAS:60477-38-5 Pifithrin-α, p-Nitro, Cyclic (PFN-α) is cell-permeable and active-form p53 inhibitor. Pifithrin-α, p-Nitro, Cyclic is one order magnitude more active than Pifithrin-α in protecting cortical neurons exposed to Etoposide (ED50=30 nM). Pifithrin-α, p-Nitro, Cyclic behaves as a p53 posttranscriptional activity inhibitor. Pifithrin-α, p-Nitro, Cyclic do not prevent p53 phosphorylation on the S15 residue[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9950