Learn More
Selleck Chemical LLC Pifithrin- (PFT) HBr 25mg 63208-82-2
SDP
Supplier:  Selleck Chemical LLC S292925MG
Encompass_Preferred
Pifithrin-? is an inhibitor of p53, inhibiting p53-dependent transactivation of p53-responsive genes. Pifithrin-? is also a potent agonist of the aryl hydrocarbon receptor (AhR). *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-3235