Learn More
Medchemexpress LLC Pifusertib hydrochloride | 2930090-28-9 | 99.9% | 460.96 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY19934A10MM/1ML
Encompass_Preferred
Pifusertib hydrochloride is a potent, selective, orally active allosteric Akt inhibitor that triggers anti-myeloma activities. It enhances fatal endoplasmic reticulum (ER) stress induced by proteasome inhibition, leading to apoptosis and autophagy.
  • Potent, selective, orally active allosteric Akt inhibitor.
  • Triggers anti-myeloma activities.
  • Enhances fatal endoplasmic reticulum (ER) stress.
  • Induces apoptosis and autophagy.
  • Blocks basal phosphorylation of Akt and downstream targets in MM cells.
  • Induces cytotoxicity in MM cells with high baseline Akt phosphorylation.
  • Inhibits tumor growth in murine xenograft models.
Catalog No. 50-004-01510