Learn More
Medchemexpress LLC Pifusertib hydrochloride | 2930090-28-9 | 99.9% | 460.96 | 5 MG
SDP
Catalog No. 5000401959
Encompass_Preferred
Pifusertib hydrochloride is a potent, selective, orally active allosteric Akt inhibitor. It is for research use only and not sold to patients. It triggers anti-myeloma activities and enhances fatal endoplasmic reticulum (ER) stress induced by proteasome inhibition.
  • Induces apoptosis and autophagy
  • Blocks basal phosphorylation of Akt
  • Selectively inhibits Akt
  • Triggers G0/G1 arrest
  • Enhances bortezomib-induced cytotoxicity
Catalog No. 50-004-01959 Supplier Medchemexpress LLC Supplier No. HY199345MG
Pifusertib hydrochloride is a potent, selective, orally active allosteric Akt inhibitor. It is for research use only and not sold to patients. It triggers anti-myeloma activities and enhances fatal endoplasmic reticulum (ER) stress induced by proteasome inhibition.
  • Induces apoptosis and autophagy
  • Blocks basal phosphorylation of Akt
  • Selectively inhibits Akt
  • Triggers G0/G1 arrest
  • Enhances bortezomib-induced cytotoxicity