Learn More
Medchemexpress LLC Pifusertib (TAS-117) | 1402602-94-1 | 98.7% | 424.49 g·mol⁻¹ | C26H24N4O2 | 10 MG
SDP
Catalog No. 5000297198
Encompass_Preferred
Pifusertib (TAS-117) is a potent, selective, orally active allosteric Akt inhibitor provided for research use. Supplied in milligram-scale packs, it is intended for preclinical biochemical and cellular studies probing Akt signaling. The compound includes specified physicochemical identifiers and high-performance liquid chromatography data for reproducible experimental application.
  • Potent, selective allosteric Akt inhibitor.
  • Orally active, suitable for in vivo preclinical models.
  • High purity: 98.7% (HPLC) for analytical consistency.
  • Molecular weight 424.49 g·mol⁻¹; formula C26H24N4O2.
  • Available in milligram-scale packs for laboratory use.
  • For research use only; not for human or clinical use.
Catalog No. 50-002-97198 Supplier Medchemexpress LLC Supplier No. HY1993410MG
Pifusertib (TAS-117) is a potent, selective, orally active allosteric Akt inhibitor provided for research use. Supplied in milligram-scale packs, it is intended for preclinical biochemical and cellular studies probing Akt signaling. The compound includes specified physicochemical identifiers and high-performance liquid chromatography data for reproducible experimental application.
  • Potent, selective allosteric Akt inhibitor.
  • Orally active, suitable for in vivo preclinical models.
  • High purity: 98.7% (HPLC) for analytical consistency.
  • Molecular weight 424.49 g·mol⁻¹; formula C26H24N4O2.
  • Available in milligram-scale packs for laboratory use.
  • For research use only; not for human or clinical use.