Learn More
American Research Products Inc Pig adrenocorticotropic hormon
Catalog No. 5000329954
Encompass
Pig adrenocorticotropic hormone (ACTH) ELISA kit from CUSABIO Detection Range 6 25 pg/mL-100 pg/mL Sensitivity 6 25 pg/mL Sample serum plasma tissue homogenates Method Competitive
Catalog No. 50-003-29954 Supplier American Research Products Inc Supplier No. CSBE12944P
Pig adrenocorticotropic hormone (ACTH) ELISA kit from CUSABIO Detection Range 6 25 pg/mL-100 pg/mL Sensitivity 6 25 pg/mL Sample serum plasma tissue homogenates Method Competitive