Learn More
American Research Products Inc Pig anti Mullerian hormone AM
Catalog No. 5000327463
Encompass
Pig anti-Mullerian hormone (AMH) ELISA kit from CUSABIO Detection Range 1 25 ng/mL-50 ng/mL Sensitivity 1 25 ng/mL Sample serum plasma tissue homogenates Method Competitive
Catalog No. 50-003-27463 Supplier American Research Products Inc Supplier No. CSBE13408P
Pig anti-Mullerian hormone (AMH) ELISA kit from CUSABIO Detection Range 1 25 ng/mL-50 ng/mL Sensitivity 1 25 ng/mL Sample serum plasma tissue homogenates Method Competitive