Learn More
American Research Products Inc Pig apolipoprotein A1 Apo A1
Catalog No. 5000329826
Encompass
Pig apolipoprotein A1 (Apo-A1) ELISA Kit from CUSABIO Detection Range 80 g/mL-2000 g/mL Sensitivity 50 g/mL Sample serum plasma tissue homogenates Method Competitive
Catalog No. 50-003-29826 Supplier American Research Products Inc Supplier No. CSBE06834P
Pig apolipoprotein A1 (Apo-A1) ELISA Kit from CUSABIO Detection Range 80 g/mL-2000 g/mL Sensitivity 50 g/mL Sample serum plasma tissue homogenates Method Competitive