Learn More
Biomatik Corporation Pig Apolipoprotein A1 (APOA1) ELISA Kit
Supplier:  Biomatik Corporation EKU02482
Encompass_Preferred
Pig Apolipoprotein A1 (APOA1) ELISA Kit for detecting APOA1 in serum, plasma, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 28.12-1800ng/ml. Sensitivity: 10.3ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P18648. Gene ID: 397691. Target Synonyms: Apo-A1; ApoA-1 Milano; ProapoA-I; Proapolipoprotein A-I; Truncated apolipoprotein A-I. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9128