Learn More
American Research Products Inc Pig apolipoprotein E Apo E E
Catalog No. 5000328366
Encompass
Pig apolipoprotein E (Apo-E) ELISA Kit from CUSABIO Detection Range 31 25 ng/mL-2000 ng/mL Sensitivity 7 81 ng/mL Sample serum plasma tissue homogenates Method Competitive
Catalog No. 50-003-28366 Supplier American Research Products Inc Supplier No. CSBE17889P
Pig apolipoprotein E (Apo-E) ELISA Kit from CUSABIO Detection Range 31 25 ng/mL-2000 ng/mL Sensitivity 7 81 ng/mL Sample serum plasma tissue homogenates Method Competitive