Learn More
American Research Products Inc Pig basic fibroblast growth fa
Catalog No. 5000326640
Encompass
Pig basic fibroblast growth factor bFGF ELISA Kit from CUSABIO Detection Range 4 7 pg/mL-300 pg/mL Sensitivity 1 2 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26640 Supplier American Research Products Inc Supplier No. CSBE12841P
Pig basic fibroblast growth factor bFGF ELISA Kit from CUSABIO Detection Range 4 7 pg/mL-300 pg/mL Sensitivity 1 2 pg/mL Sample serum plasma tissue homogenates Method Sandwich