Learn More
Biomatik Corporation Pig B-Cell Activating Factor (BAFF) ELISA Kit
Supplier:  Biomatik Corporation EKU10676
Encompass_Preferred
Pig B-Cell Activating Factor (BAFF) ELISA Kit for detecting BAFF in serum, plasma, tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: A0A8D1TQZ4. Gene ID: 100038026. Target Synonyms: CD257; TNFSF13B; BLYS; TALL1; THANK; TNFSF20; ZTNF4; B lymphocyte stimulator; Tumor Necrosis Factor Ligand Superfamily Member 13B; Dendritic cell-derived TNF-like. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5015