Learn More
Biomatik Corporation Pig Bone Morphogenetic Protein 2 (BMP2) CLIA Kit
Supplier:  Biomatik Corporation EKU09529
Encompass_Preferred
Pig Bone Morphogenetic Protein 2 (BMP2) CLIA Kit for detecting BMP2 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 2.74-2,000pg/ml. Sensitivity: 1.13pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: E1AXT2. Gene ID: 100157103. Target Synonyms: BMP2A; BMP-2A; Hemochromatosis Modifier. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8503