Learn More
Biomatik Corporation Pig Chemerin (CHEM) ELISA Kit
Supplier:  Biomatik Corporation EKU11687
Encompass_Preferred
Pig Chemerin (CHEM) ELISA Kit for detecting CHEM in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: B0LUW3. Gene ID: 100141314. Target Synonyms: TIG2; RARRES2; Tazarotene-Induced Gene 2; Retinoic Acid Receptor Responder 2,Tazarotene Induced; RAR-responsive protein TIG2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6184